Anti-FOXO1 Rabbit Polyclonal Antibody
Catalog # 77699-531
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FOXO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q12778
- Antigen Synonyms:Drosophila|FKHR|Forkhead in rhabdomyosarcoma|FOXO1A|foxo1|Forkhead box protein O1|Forkhead|FoxO transcription factor|FKH 1|Forkhead box O1|Forkhead (Drosophila) homolog 1 (rhabdomyosarcoma)|Forkhead box protein O1A|homolog of|OTTHUMP00000018301|FOXO1_HUMAN|FKH1|in rhabdomyosarcoma
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human FoxO1 around the non-phosphorylation site of S319.
- Purification:Antigen affinity purified
- Cat. No.:77699-531
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: FOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat