Anti-EID1 Rabbit Polyclonal Antibody
77699-419
New Product
- Antigen Symbol:EID1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9Y6B2
- Antigen Synonyms:Rb and p300 binding protein EID 1|CBP|MGC138884|IRO45620|EID1_HUMAN|NB4 apoptosis related protein |PTD014|EID-1|MGC138883|Rb and p300 binding protein EID1 |RBP21|Retinoblastoma protein associated protein|C15orf3|21 kDa pRb associated protein |CREBBP/EP300 inhibitory protein 1|EP300-interacting inhibitor of differentiation 1|21 kDa pRb-associated protein|EID1|EID 1|EP300 interacting inhibitor of differentiation 1|PNAS22 |E1A like inhibitor of differentiation|Chromosome 15 open reading frame 3 |PNAS 22|E1A-like inhibitor of differentiation 1|CRI 1|CREBBP/EP300 inhibitor 1|CRI1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human EID-1.
- Purification:Antigen affinity purified
- Cat. No.:77699-419
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: EID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human