Anti-SRC Rabbit Polyclonal Antibody
77699-244
New Product
- Antigen Symbol:SRC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P12931
- Antigen Synonyms:c SRC|ASV|Oncogene SRC|p60c-src|Neuronal SRC|Src|SRC proto oncogene non receptor tyrosine kinase|SRC_HUMAN|pp60c src|p60 Src|Tyrosine protein kinase SRC 1|SRC Oncogene|p60-Src|Tyrosine kinase pp60c src|pp60csrc|Neuronal proto-oncogene tyrosine-protein kinase Src|Protooncogene SRC Rous sarcoma|Proto-oncogene c-Src|p60Src|AW259666|CDNA FLJ14219 fis clone NT2RP3003800 highly similar to Rattus norvegicus tyrosine protein kinase pp60 c src mRNA|Protooncogene SRC|v src sarcoma (Schmidt Ruppin A 2) viral oncogene homolog avian|Neuronal CSRC tyrosine specific protein kinase|OTTHUMP00000174476|Avian sarcoma virus|v src avian sarcoma (Schmidt Ruppin A2) viral oncogene homolog|pp60c-src|SRC1|Proto oncogene tyrosine protein kinase Src|OTTHUMP00000174477|Proto-oncogene tyrosine-protein kinase Src|Tyrosine protein kinase SRC1|V src sarcoma (Schmidt Ruppin A 2) viral oncogene homolog (avian)|EC 2.7.10.2|cSrc
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human c-Src around the non-phosphorylation site of Y419.
- Purification:Antigen affinity purified
- Cat. No.:77699-244
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: SRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat