Anti-CREB3L2 Rabbit Polyclonal Antibody
Catalog # 77699-230
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CREB3L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q70SY1
- Antigen Synonyms:cAMP responsive element binding protein 3 like 2|BBF2H7|BZIB transcription factor|cAMP-responsive element-binding protein 3-like protein 2|Creb3l2|Basic transcription factor 2|FUS/BBF2H7 protein |TCAG_1951439|Processed cyclic AMP-responsive element-binding protein 3-like protein 2|Cyclic AMP-responsive element-binding protein 3-like protein 2|BBF2 human homolog on chromosome 7|CR3L2_HUMAN|B ZIB transcription factor|MGC131709|MGC71006
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CREB3L2.
- Purification:Antigen affinity purified
- Cat. No.:77699-230
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: CREB3L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat