Anti-TFAP2C Rabbit Polyclonal Antibody
Catalog # 77698-864
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:TFAP2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q92754
- Antigen Synonyms:Activating enhancer-binding protein 2 gamma|Estrogen receptor factor 1|hAP 2g|ERF 1|Transcription factor ERF1|AP2g|Transcription factor ERF-1|Transcription factor AP 2 gamma|Transcription factor AP 2 gamma (activating enhancer binding protein 2 gamma)|hAP2g|Activating enhancer binding protein 2 gamma|TFAP2G|Transcription factor ERF 1|ERF1|AP2C_HUMAN|Transcription factor AP-2 gamma|AP2-gamma|AP2gamma|Tfap2c|TFAP 2C|TFAP 2G
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human AP-2γ.
- Purification:Antigen affinity purified
- Cat. No.:77698-864
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: TFAP2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat