Anti-TMEM30CP Rabbit Polyclonal Antibody
Catalog # 77702-682
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TMEM30CP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:A0ZSE6
- Antigen Synonyms:TMEM30CP|TMEM30C|CDC50C|Cell cycle control protein 50C|Transmembrane protein 30C
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Cdc50C.
- Purification:Antigen affinity purified
- Cat. No.:77702-682
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: TMEM30CP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human