Anti-PBOV1 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:10-1:50
Type:
Antigen: PBOV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77712-658
- 77712-657
- Antigen Symbol:
- PBOV1
- PBOV1
- Antigen Synonyms:
- Prostate and breast cancer overexpressed 1|PBOV1|UROC28|UC28|Prostate and breast cancer overexpressed gene 1 protein|Protein UROC28|ibody|PBOV1_HUMAN
- Prostate and breast cancer overexpressed 1|PBOV1|UROC28|UC28|Prostate and breast cancer overexpressed gene 1 protein|Protein UROC28|ibody|PBOV1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZY1
- Q9GZY1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PBOV1
- Synthetic peptide of Human PBOV1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice