Anti-LY6E Rabbit Polyclonal Antibody
Catalog # 77703-071
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LY6E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q16553
- Antigen Synonyms:Retinoic acid-induced gene E protein|TSA1|RIGE|SCA-2|9804|Ly-6E|RIG-E|Thymic shared antigen 1|Stem cell antigen 2|Lymphocyte antigen 6E|SCA2|LY6E|TSA-1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human TSA-1.
- Purification:Antigen affinity purified
- Cat. No.:77703-071
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: LY6E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human