Anti-GLI3 Rabbit Polyclonal Antibody
Catalog # 77702-616
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GLI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P10071
- Antigen Synonyms:PAPA 1|Oncogene GLI3|GLI3 form of 83 kDa|Zinc finger protein GLI3|GLI3_HUMAN|PAPA|GLI3-83|GLI3 C-terminally truncated form|GLI3 form of 190 kDa|Glioma associated oncogene family zinc finger 3|GLI3FL|GCPS|Transcriptional activator GLI3|GLI Kruppel family member GLI 3|ACLS|PHS|Gli 3|GLI3|PAPB|DNA binding protein|GLI3-190|GLI Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome)|Zinc finger protein GLI 3|GLI family zinc finger 3|PPDIV|GLI Kruppel family member GLI3|PAP A|Transcriptional repressor GLI3R|PAPA1|GLI3 full length protein|PPD IV
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human GLI-3.
- Purification:Antigen affinity purified
- Cat. No.:77702-616
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: GLI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat