Anti-GAP43 (phospho-S41) Rabbit Polyclonal Antibody
Catalog # 77702-511
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GAP43 (phospho-S41)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P17677
- Antigen Synonyms:QtrA-11580|Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin binding protein P 57|Axonal membrane protein GAP 43|Nerve growth related peptide GAP43|Growth Associated Protein 43|Nerve Growth Related Peptide|Neural phosphoprotein B-50|QtrA-13071|NEUM_HUMAN|Protein F1|Neural phosphoprotein B 50|GAP43|Neuromodulin|Neuron growth associated protein 43|GAP 43|B 50|F1|PP46
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human GAP-43 around the phosphorylation site of S41.
- Purification:Antigen affinity purified
- Cat. No.:77702-511
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: GAP43 (phospho-S41)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat