Anti-GABRA1 Rabbit Polyclonal Antibody
Catalog # 77702-483
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GABRA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P14867
- Antigen Synonyms:GABA(A) receptor subunit alpha-1|Gamma aminobutyric acid (GABA) A receptor alpha 1|Gamma aminobutyric acid type A receptor alpha1 subunit|GABA(A) receptor|Gamma-aminobutyric acid receptor subunit alpha-1|GABRA1|ECA4|EIEE19|Gamma aminobutyric acid receptor subunit alpha 1|Gaba receptor alpha 1 polypeptide|GBRA1_HUMAN|EJM5|GABA(A) receptor, alpha 1|GABRA 1|Gamma aminobutyric acid A receptor alpha 1|GABA(A) receptor subunit alpha 1|EJM
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human GABAA Rα1.
- Purification:Antigen affinity purified
- Cat. No.:77702-483
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: GABRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat