Anti-COX5A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: COX5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77712-465
- 77712-466
- Antigen Symbol:
- COX5A
- COX5A
- Antigen Synonyms:
- COX5A_HUMAN|Cytochrome c oxidase subunit 5A|COX5A|VA|COX|Cytochrome c oxidase polypeptide|Cytochrome c oxidase subunit Va|COX VA|mitochondrial|Mitochondrial cytochrome c oxidase subunit Va|Cytochrome c oxidase polypeptide Va
- COX5A_HUMAN|Cytochrome c oxidase subunit 5A|COX5A|VA|COX|Cytochrome c oxidase polypeptide|Cytochrome c oxidase subunit Va|COX VA|mitochondrial|Mitochondrial cytochrome c oxidase subunit Va|Cytochrome c oxidase polypeptide Va
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20674
- P20674
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human COX5A
- Synthetic peptide of Human COX5A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice