Anti-AQP6 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AQP6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77710-950
- 77710-949
- Antigen Symbol:
- AQP6
- AQP6
- Antigen Synonyms:
- AQP6_HUMAN|Aquaporin 6|Aqp6|Kidney-specific aquaporin|Human kidney water channel (hKID) mRNA|AQP2L|Aquaporin-6|complete cds|aquaporin 2 like kidney specific|KID|Aquaporin 6 kidney specific|Aquaporin 2 like|Aquaporin-2-like|AQP-6|hKID
- AQP6_HUMAN|Aquaporin 6|Aqp6|Kidney-specific aquaporin|Human kidney water channel (hKID) mRNA|AQP2L|Aquaporin-6|complete cds|aquaporin 2 like kidney specific|KID|Aquaporin 6 kidney specific|Aquaporin 2 like|Aquaporin-2-like|AQP-6|hKID
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13520
- Q13520
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Aquaporin-6 protein (1-30AA)
- Recombinant Human Aquaporin-6 protein (1-30AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice