Anti-SFR1 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SFR1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77716-690
- 77716-689
- Antigen Symbol:
- SFR1
- SFR1
- Antigen Synonyms:
- Meiosis protein 5 homolog|SFR1|MEI5|Swi5-dependent recombination DNA repair protein 1 homolog|C10orf78|MEIR5
- Meiosis protein 5 homolog|SFR1|MEI5|Swi5-dependent recombination DNA repair protein 1 homolog|C10orf78|MEIR5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86XK3
- Q86XK3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Swi5-dependent recombination DNA repair protein 1 homolog protein (5-93AA)
- Recombinant Human Swi5-dependent recombination DNA repair protein 1 homolog protein (5-93AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice