Anti-SGMS1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SGMS1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77716-654
- 77716-653
- Antigen Symbol:
- SGMS1
- SGMS1
- Antigen Synonyms:
- SGMS1|Protein Mob|Sphingomyelin synthase 1|SMS1|Phosphatidylcholine:ceramide cholinephosphotransferase 1|Transmembrane protein 23|Medulla oblongata-derived protein|EC 2.7.8.27|TMEM23|MOB
- SGMS1|Protein Mob|Sphingomyelin synthase 1|SMS1|Phosphatidylcholine:ceramide cholinephosphotransferase 1|Transmembrane protein 23|Medulla oblongata-derived protein|EC 2.7.8.27|TMEM23|MOB
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86VZ5
- Q86VZ5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Phosphatidylcholine:ceramide cholinephosphotransferase 1 protein (48-137AA)
- Recombinant Human Phosphatidylcholine:ceramide cholinephosphotransferase 1 protein (48-137AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice