Anti-PSMD3 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: PSMD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77715-584
- 77715-583
- Antigen Symbol:
- PSMD3
- PSMD3
- Antigen Synonyms:
- 26S proteasome regulatory subunit S3|26S proteasome regulatory subunit RPN3|Proteasome subunit p58|PSMD3_HUMAN|26S proteasome non-ATPase regulatory subunit 3|RPN3|Tissue specific transplantation antigen 2|PSMD3|Proteasome (prosome, macropain) 26S subunit non ATPase 3|TSTA2|26S proteasome non ATPase regulatory subunit 3|S3|OTTHUMP00000164347|P58
- 26S proteasome regulatory subunit S3|26S proteasome regulatory subunit RPN3|Proteasome subunit p58|PSMD3_HUMAN|26S proteasome non-ATPase regulatory subunit 3|RPN3|Tissue specific transplantation antigen 2|PSMD3|Proteasome (prosome, macropain) 26S subunit non ATPase 3|TSTA2|26S proteasome non ATPase regulatory subunit 3|S3|OTTHUMP00000164347|P58
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43242
- O43242
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PSMD3
- Fusion protein of Human PSMD3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice