Anti-AP1G1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:25-1:100
Type:
Antigen: AP1G1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77715-436
- 77715-437
- Antigen Symbol:
- AP1G1
- AP1G1
- Antigen Synonyms:
- Adaptor related protein complex 1 gamma 1 subunit|Clathrin associated/assembly/adaptor protein large gamma 1|AP1G1_HUMAN|CLAPG1|Adaptor protein complex AP 1 subunit gamma 1|Adaptor protein complex AP-1 subunit gamma-1|Golgi adaptor HA1/AP1 adaptin subunit gamma 1|Clathrin assembly protein complex 1 gamma 1 large chain|AP1G1|Golgi adaptor HA1/AP1 adaptin subunit gamma-1|Gamma1 adaptin|Adapter-related protein complex 1 subunit gamma-1|AP 1 complex subunit gamma 1|MGC18255|ADTG|Gamma1-adaptin|AP-1 complex subunit gamma-1|Golgi adaptor HA1/AP1 adaptin gamma subunit|Clathrin assembly protein complex 1 gamma large chain|Adaptor protein complex AP 1 gamma 1 subunit|Clathrin assembly protein complex 1 gamma-1 large chain
- Adaptor related protein complex 1 gamma 1 subunit|Clathrin associated/assembly/adaptor protein large gamma 1|AP1G1_HUMAN|CLAPG1|Adaptor protein complex AP 1 subunit gamma 1|Adaptor protein complex AP-1 subunit gamma-1|Golgi adaptor HA1/AP1 adaptin subunit gamma 1|Clathrin assembly protein complex 1 gamma 1 large chain|AP1G1|Golgi adaptor HA1/AP1 adaptin subunit gamma-1|Gamma1 adaptin|Adapter-related protein complex 1 subunit gamma-1|AP 1 complex subunit gamma 1|MGC18255|ADTG|Gamma1-adaptin|AP-1 complex subunit gamma-1|Golgi adaptor HA1/AP1 adaptin gamma subunit|Clathrin assembly protein complex 1 gamma large chain|Adaptor protein complex AP 1 gamma 1 subunit|Clathrin assembly protein complex 1 gamma-1 large chain
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43747
- O43747
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human AP1G1
- Synthetic peptide of Human AP1G1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice