Anti-PPP1R3A Rabbit Polyclonal Antibody
Catalog # 77703-767
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:PPP1R3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q16821
- Antigen Synonyms:Protein phosphatase 1, regulatory subunit 3|protein phosphatase 1, regulatory (inhibitor) subunit 3A|PPP1R3|PPR3A_HUMAN|GM|Glycogen associated regulatory subunit of protein phosphatase 1|Protein phosphatase 1 regulatory subunit 3A|Type 1 protein phosphatase skeletal muscle glycogen targeting subunit|Glycogen and sarcoplasmic reticulum binding subunit, skeletal muscle|Protein phosphatase 1 glycogen-associated regulatory subunit|Serine /threonine specific protein phosphatase|RG1|Protein phosphatase 1 glycogen-targeting subunit, muscle|PP1G|PPP1R3A|Protein phosphatase 1 regulatory subunit GM|Protein phosphatase type-1 glycogen targeting subunit
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human PPP1R3A.
- Purification:Antigen affinity purified
- Cat. No.:77703-767
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PPP1R3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human