Anti-CSNK1G1/CSNK1G2/CSNK1G3 Rabbit Polyclonal Antibody
Catalog # 77703-699
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CSNK1G1/CSNK1G2/CSNK1G3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9HCP0/P78368/Q9Y6M4
- Antigen Synonyms:Casein kinase I isoform gamma-1|Casein kinase I isoform gamma-3|Casein kinase I isoform gamma-2|CSNK1G1|CSNK1G3|CKI-gamma 2|CK1G2|CKI-gamma 3|CKI-gamma 1|CSNK1G2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Casein Kinase Iγ1/2/3 around the non-phosphorylation site of Y263.
- Purification:Antigen affinity purified
- Cat. No.:77703-699
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: CSNK1G1/CSNK1G2/CSNK1G3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat