Anti-TOX3 Rabbit Polyclonal Antibody
77688-817
New Product
- Antigen Symbol:TOX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:O15405
- Antigen Synonyms:CAG trinucleotide repeat-containing gene F9 protein|CAGF9|TOX3|Trinucleotide repeat containing gene 9 protein|TOX high mobility group box family member 3|BC052044|Trinucleotide repeat containing 9|TOX3_HUMAN|TNRC9|C230068E13
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human TOX high mobility group box family member 3 protein (8-305AA)
- Purification:>95%, Protein G purified
- Cat. No.:77688-817
- Packaging:Vial
Type:
Antigen: TOX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse