To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:RAF1 (phospho-S259)
- Clonality:Recombinant Monoclonal
- Clone:4C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P04049
- Antigen Synonyms:Raf-1|RAF1|RAF|cRaf|RAF proto-oncogene serine/threonine-protein kinase|Proto-oncogene c-RAF
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-RAF1 (S259)
- Purification:Affinity purified
- Cat. No.:77691-337
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:50-1:200, IF:1:20-1:200
Type:
Antigen: RAF1 (phospho-S259)
Clonality: Recombinant Monoclonal
Clone: 4C9
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human