To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:Histone H4 (acetyl-K16)
- Clonality:Recombinant Monoclonal
- Clone:2B8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P62805
- Antigen Synonyms:H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|Histone H4|HIST1H4B|H4/G|HIST2H4B|HIST1H4E|H4F2|H4/O|HIST1H4L|H4/D|HIST4H4|H4FC|AND|H4/E|H4/B|H4FE|HIST1H4A|HIST1H4J|H4FH|H4FM|H4FO|H4FB|HIST1H4K|H4/N|H4/J|HIST2H4|H4/C|HIST1H4F|HIST1H4D|H4FG|HIST1H4I|H4/A|H4FK|H4/H|HIST1H4C|H4FN|H4FD|HIST2H4A
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Cat. No.:77691-291
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, ICC:1:50-1:500, IF:1:50-1:200
Type:
Antigen: Histone H4 (acetyl-K16)
Clonality: Recombinant Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human, Mouse