To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:HIST1H2AB
- Clonality:Recombinant Monoclonal
- Clone:1H12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P04908
- Antigen Synonyms:AND|H2AFA|HIST1H2AB|Histone H2A/a|HIST1H2AE|Histone H2A.2|Histone H2A type 1-B/E|H2AFM|Histone H2A/m
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Cat. No.:77691-273
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IF:1:30-1:200
Type:
Antigen: HIST1H2AB
Clonality: Recombinant Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human