Anti-PNPLA3 Rabbit Polyclonal Antibody
77691-024
New Product
- Antigen Symbol:PNPLA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NST1
- Antigen Synonyms:Lysophosphatidic acid acyltransferase|EC 3.1.1.3|Acylglycerol transacylase|Adiponutrin|C22orf20|PNPLA3|Patatin-like phospholipase domain-containing protein 3|1-acylglycerol-3-phosphate O-acyltransferase PNPLA3|EC 2.3.1.51|ADPN|Calcium-independent phospholipase A2-epsilon|iPLA2-epsilon
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Patatin-like phospholipase domain-containing protein 3 protein (377-481AA)
- Purification:Antigen affinity purified
- Cat. No.:77691-024
- Packaging:Vial
Recommended dilution: WB:1:200-1:1000, IHC:1:20-1:200
Type:
Antigen: PNPLA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human