Anti-ABCB9 Rabbit Polyclonal Antibody
77691-019
New Product
- Antigen Symbol:ABCB9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NP78
- Antigen Synonyms:ATP-binding cassette transporter 9|TAP-like protein|ABCB9|hABCB9|TAPL|KIAA1520ATP-binding cassette sub-family B member 9|ABC transporter 9 protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP-binding cassette sub-family B member 9 protein (630-766AA)
- Purification:>95%, Protein G purified
- Cat. No.:77691-019
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200
Type:
Antigen: ABCB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse