Anti-NOP53 Rabbit Polyclonal Antibody
Catalog # 77690-974
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NOP53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NZM5
- Antigen Synonyms:GSCR2_HUMAN|GLTSCR2|p60|PICT1|protein interacting with carboxyl terminus 1|Glioma tumor suppressor candidate region gene 2 protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ribosome biogenesis protein NOP53 protein (227-405AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-974
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NOP53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat