Anti-GFM2 Rabbit Polyclonal Antibody
Catalog # 77690-505
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GFM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q969S9
- Antigen Synonyms:mEF-G 2|RRF2mt|Elongation factor G 2|hEFG2|Ribosome-releasing factor 2|mitochondrial|EFG2|MSTP027|Elongation factor G2|EF-G2mt|GFM2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ribosome-releasing factor 2, mitochondrial protein (520-779AA)
- Purification:Antigen affinity purified
- Cat. No.:77690-505
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: GFM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human