Anti-RABGGTA Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: RABGGTA
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77725-606
- 77725-607
- Antigen Symbol:
- RABGGTA
- RABGGTA
- Antigen Synonyms:
- PGTA_HUMAN|Geranylgeranyl transferase type 2 subunit alpha|Rab GG transferase alpha|Rab geranyl-geranyltransferase subunit alpha|Geranylgeranyl transferase type II subunit alpha|Rab geranylgeranyltransferase subunit alpha|RABGGTA|Rab geranyl geranyltransferase subunit alpha|Geranylgeranyl transferase type-2 subunit alpha|Rab GGTase alpha|Rab geranylgeranyltransferase alpha subunit|PTAR3|Protein prenyltransferase alpha subunit repeat containing 3
- PGTA_HUMAN|Geranylgeranyl transferase type 2 subunit alpha|Rab GG transferase alpha|Rab geranyl-geranyltransferase subunit alpha|Geranylgeranyl transferase type II subunit alpha|Rab geranylgeranyltransferase subunit alpha|RABGGTA|Rab geranyl geranyltransferase subunit alpha|Geranylgeranyl transferase type-2 subunit alpha|Rab GGTase alpha|Rab geranylgeranyltransferase alpha subunit|PTAR3|Protein prenyltransferase alpha subunit repeat containing 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92696
- Q92696
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Geranylgeranyl transferase type-2 subunit alpha protein (1-200AA)
- Recombinant Human Geranylgeranyl transferase type-2 subunit alpha protein (1-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice