Anti-ascl1a Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ascl1a
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Specifications
- Catalog No:
- 77725-536
- 77725-535
- Antigen Symbol:
- ascl1a
- ascl1a
- Antigen Synonyms:
- Zash-1a|ash|ash1a|ascl1a|Pituitary-absent protein|asha|piaAchaete-scute homolog 1a
- Zash-1a|ash|ash1a|ascl1a|Pituitary-absent protein|asha|piaAchaete-scute homolog 1a
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Zebrafish
- Zebrafish
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q90259
- Q90259
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Zebrafish Achaete-scute homolog 1a protein (1-196AA)
- Recombinant Zebrafish Achaete-scute homolog 1a protein (1-196AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice