Anti-STK35 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STK35
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-404
- 77725-405
- Antigen Symbol:
- STK35
- STK35
- Antigen Synonyms:
- Serine/threonine-protein kinase 35 L1|CLP-36-interacting kinase 1|PDLIM1-interacting kinase 1|Serine threonine kinase 35 long form|CLP 36 interacting kinase|STK35L1|Serine/threonine kinase 35|Clik1|PDLIM1 interacting kinase|STK35_HUMAN|Serine/threonine-protein kinase 35|Serine/threonine protein kinase 35|CLIK-1|STK35|EC 2.7.1.37
- Serine/threonine-protein kinase 35 L1|CLP-36-interacting kinase 1|PDLIM1-interacting kinase 1|Serine threonine kinase 35 long form|CLP 36 interacting kinase|STK35L1|Serine/threonine kinase 35|Clik1|PDLIM1 interacting kinase|STK35_HUMAN|Serine/threonine-protein kinase 35|Serine/threonine protein kinase 35|CLIK-1|STK35|EC 2.7.1.37
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDR2
- Q8TDR2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase 35 protein (1-200AA)
- Recombinant Human Serine/threonine-protein kinase 35 protein (1-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice