Anti-TDRD9 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: TDRD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-281
- 77690-440
- Antigen Symbol:
- TDRD9
- TDRD9
- Antigen Synonyms:
- HIG 1|FLJ36164|TDRD 9|Tdrd9|Tudor domain containing protein 9|Putative ATP dependent RNA helicase TDRD9|TDRD9_HUMAN|MGC135025|Putative ATP-dependent RNA helicase TDRD9|Tudor domain-containing protein 9|Tudor domain containing 9|C14orf75|DKFZp434N0820|NET54|Hypoxia inducible HIG 1|chromosome 14 open reading frame 75
- HIG 1|FLJ36164|TDRD 9|Tdrd9|Tudor domain containing protein 9|Putative ATP dependent RNA helicase TDRD9|TDRD9_HUMAN|MGC135025|Putative ATP-dependent RNA helicase TDRD9|Tudor domain-containing protein 9|Tudor domain containing 9|C14orf75|DKFZp434N0820|NET54|Hypoxia inducible HIG 1|chromosome 14 open reading frame 75
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NDG6
- Q8NDG6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Putative ATP-dependent RNA helicase TDRD9 protein (286-585AA)
- Recombinant Human Putative ATP-dependent RNA helicase TDRD9 protein (286-585AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice