Anti-SLAIN1 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SLAIN1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77725-275
- 77725-276
- Antigen Symbol:
- SLAIN1
- SLAIN1
- Antigen Synonyms:
- FLJ30046|Slain1|member 1|MGC131899|SLAIN motif-containing protein 1|chromosome 13 open reading frame 32|SLAIN motif family member 1|SLAI1_HUMAN|C13orf32|SLAIN motif family
- FLJ30046|Slain1|member 1|MGC131899|SLAIN motif-containing protein 1|chromosome 13 open reading frame 32|SLAIN motif family member 1|SLAI1_HUMAN|C13orf32|SLAIN motif family
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8ND83
- Q8ND83
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human SLAIN motif-containing protein 1 protein (43-136AA)
- Recombinant Human SLAIN motif-containing protein 1 protein (43-136AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice