Anti-NGEF Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NGEF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-164
- 77725-165
- Antigen Symbol:
- NGEF
- NGEF
- Antigen Synonyms:
- Ephexin-1|NGEF_HUMAN|Ngef|ARHGEF27|Eph-interacting exchange protein|EPHEXIN|Guanine nucleotide exchange factor, neuronal|Neuronal guanine nucleotide exchange factor|Eph receptor-interacting exchange protein
- Ephexin-1|NGEF_HUMAN|Ngef|ARHGEF27|Eph-interacting exchange protein|EPHEXIN|Guanine nucleotide exchange factor, neuronal|Neuronal guanine nucleotide exchange factor|Eph receptor-interacting exchange protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N5V2
- Q8N5V2
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Ephexin-1 protein (495-710AA)
- Recombinant Human Ephexin-1 protein (495-710AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice