Anti-UBL7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: UBL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-039
- 77725-038
- Antigen Symbol:
- UBL7
- UBL7
- Antigen Synonyms:
- BMSCUBP|Ubiquitin like 7|Bone marrow stromal cell ubiquitin-like protein|Ubl7|Ubiquitin-like protein 7|Bone marrow stromal cell derived ubiquitin like protein|BMSC-UbP|2300004C15Rik|Ubiquitin like protein SB132|SB132|BMSC UbP|Ubiquitin-like protein SB132|TCBA1|Ubiquitin like 7 (bone marrow stromal cell derived)|UBL7_HUMAN|MGC94284|MGC14421
- BMSCUBP|Ubiquitin like 7|Bone marrow stromal cell ubiquitin-like protein|Ubl7|Ubiquitin-like protein 7|Bone marrow stromal cell derived ubiquitin like protein|BMSC-UbP|2300004C15Rik|Ubiquitin like protein SB132|SB132|BMSC UbP|Ubiquitin-like protein SB132|TCBA1|Ubiquitin like 7 (bone marrow stromal cell derived)|UBL7_HUMAN|MGC94284|MGC14421
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96S82
- Q96S82
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-like protein 7 protein (1-300AA)
- Recombinant Human Ubiquitin-like protein 7 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice