Anti-MRGPRX3 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRGPRX3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-921
- 77724-922
- Antigen Symbol:
- MRGPRX3
- MRGPRX3
- Antigen Synonyms:
- G protein coupled receptor SNSR2|MAS-related GPR, member X3|Mas-related G-protein coupled receptor member X3|G protein coupled receptor SNSR1|MRGX3_HUMAN|Mas-related G protein coupled MRGX3|Mas-related G protein coupled MRGX 3|MRGX3|GPCR|SNSR1|G protein-coupled receptor MRGX 3|SNSR2|Mas related GPCR member X3|Sensory neuron-specific G-protein coupled receptor 1|Sensory neuron-specific G-protein coupled receptor 1/2|MRGPRX3|G protein-coupled receptor MRGX3
- G protein coupled receptor SNSR2|MAS-related GPR, member X3|Mas-related G-protein coupled receptor member X3|G protein coupled receptor SNSR1|MRGX3_HUMAN|Mas-related G protein coupled MRGX3|Mas-related G protein coupled MRGX 3|MRGX3|GPCR|SNSR1|G protein-coupled receptor MRGX 3|SNSR2|Mas related GPCR member X3|Sensory neuron-specific G-protein coupled receptor 1|Sensory neuron-specific G-protein coupled receptor 1/2|MRGPRX3|G protein-coupled receptor MRGX3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96LB0
- Q96LB0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mas-related G-protein coupled receptor member X3 protein (276-322AA)
- Recombinant Human Mas-related G-protein coupled receptor member X3 protein (276-322AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice