Anti-IL22RA2 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: IL22RA2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77724-646
- 77724-645
- Antigen Symbol:
- IL22RA2
- IL22RA2
- Antigen Synonyms:
- Cytokine receptor class II member 10|IL-22 receptor subunit alpha-2|Interleukin 22 Receptor alpha 2|IL 22R alpha 2|IL 22RA2|Cytokine receptor class-II member 10|IL 22 receptor subunit alpha 2|soluble 1|Il22ra2|I22R2_HUMAN|IL-22BP|IL-22R-alpha-2|Class II cytokine receptor|CRF2-S1|MGC150509|Interleukin-22-binding protein|IL22RA2|Cytokine receptor family 2 member 10|Cytokine receptor family type 2 soluble 1|CRF2X|IL 22BP|CRF2 10|Interleukin-22 receptor subunit alpha-2|CRF2 S1|CRF2-10|ZcytoR16|Interleukin 22 binding protein|IL-22RA2|IL22 RA2|IL22BP|Cytokine receptor family type 2|IL22R alpha 2|MGC150510
- Cytokine receptor class II member 10|IL-22 receptor subunit alpha-2|Interleukin 22 Receptor alpha 2|IL 22R alpha 2|IL 22RA2|Cytokine receptor class-II member 10|IL 22 receptor subunit alpha 2|soluble 1|Il22ra2|I22R2_HUMAN|IL-22BP|IL-22R-alpha-2|Class II cytokine receptor|CRF2-S1|MGC150509|Interleukin-22-binding protein|IL22RA2|Cytokine receptor family 2 member 10|Cytokine receptor family type 2 soluble 1|CRF2X|IL 22BP|CRF2 10|Interleukin-22 receptor subunit alpha-2|CRF2 S1|CRF2-10|ZcytoR16|Interleukin 22 binding protein|IL-22RA2|IL22 RA2|IL22BP|Cytokine receptor family type 2|IL22R alpha 2|MGC150510
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969J5
- Q969J5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Interleukin-22 receptor subunit alpha-2 protein (22-263AA)
- Recombinant Human Interleukin-22 receptor subunit alpha-2 protein (22-263AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice