Anti-LNX1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: LNX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-360
- 77724-359
- Antigen Symbol:
- LNX1
- LNX1
- Antigen Synonyms:
- Numb-binding protein 1|LNX|EC 2.3.2.27|UNQ574/PRO1136|RING-type E3 ubiquitin transferase LNX|PDZ domain-containing RING finger protein 2|LNX1|E3 ubiquitin-protein ligase LNX|Ligand of Numb-protein X 1|PDZRN2
- Numb-binding protein 1|LNX|EC 2.3.2.27|UNQ574/PRO1136|RING-type E3 ubiquitin transferase LNX|PDZ domain-containing RING finger protein 2|LNX1|E3 ubiquitin-protein ligase LNX|Ligand of Numb-protein X 1|PDZRN2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TBB1
- Q8TBB1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase LNX protein (133-259AA)
- Recombinant Human E3 ubiquitin-protein ligase LNX protein (133-259AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice