Anti-LILRB1 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LILRB1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-332
- 77724-331
- Antigen Symbol:
- LILRB1
- LILRB1
- Antigen Synonyms:
- Ig like transcript 2|PIRB|ILT-2|LIR-1|immunoglobulin like transcript 2|CD85j antigen|LIR1|leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1|CD85|LIR 1|Leukocyte immunoglobulin-like receptor 1|CD85 antigen|leukocyte immunoglobulin like receptor, subfamily B, member 1|MIR7|MIR-7|LILRB1|CD85 antigen like family member J|Leukocyte immunoglobulin like receptor 1|MIR 7|Monocyte/macrophage immunoglobulin like receptor 7|leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform|leukocyte immunoglobulin like receptor subfamily B member 1 soluble isoform|Leukocyte immunoglobulin-like receptor subfamily B member 1|ILT2|leukocyte Ig like receptor 1|Immunoglobulin-like transcript 2|leukocyte immunoglobulin like receptor subfamily B member 1|CD85j|PIR B|ILT 2|Monocyte/macrophage immunoglobulin-like receptor 7|CD85 antigen-like family member J|Leukocyte immunoglobulin like receptor subfamily B member 1 precursor|LIRB1_HUMAN
- Ig like transcript 2|PIRB|ILT-2|LIR-1|immunoglobulin like transcript 2|CD85j antigen|LIR1|leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1|CD85|LIR 1|Leukocyte immunoglobulin-like receptor 1|CD85 antigen|leukocyte immunoglobulin like receptor, subfamily B, member 1|MIR7|MIR-7|LILRB1|CD85 antigen like family member J|Leukocyte immunoglobulin like receptor 1|MIR 7|Monocyte/macrophage immunoglobulin like receptor 7|leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform|leukocyte immunoglobulin like receptor subfamily B member 1 soluble isoform|Leukocyte immunoglobulin-like receptor subfamily B member 1|ILT2|leukocyte Ig like receptor 1|Immunoglobulin-like transcript 2|leukocyte immunoglobulin like receptor subfamily B member 1|CD85j|PIR B|ILT 2|Monocyte/macrophage immunoglobulin-like receptor 7|CD85 antigen-like family member J|Leukocyte immunoglobulin like receptor subfamily B member 1 precursor|LIRB1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NHL6
- Q8NHL6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leukocyte immunoglobulin-like receptor subfamily B member 1 protein (483-650AA)
- Recombinant Human Leukocyte immunoglobulin-like receptor subfamily B member 1 protein (483-650AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice