Anti-ADSS Rabbit Polyclonal Antibody
Catalog # 77687-136
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADSS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P30520
- Antigen Synonyms:acidic isozyme|AdSS 2|Adenylosuccinate synthetase isozyme 2|IMP--aspartate ligase 2|L-type adenylosuccinate synthetase|Adenylosuccinate synthetase|liver isozyme|AMPSase 2|ADSS|ADSS2|EC 6.3.4.4
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenylosuccinate synthetase isozyme 2 protein (1-280AA)
- Purification:Antigen affinity purified
- Cat. No.:77687-136
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: ADSS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse