To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:CAMK2A/CAMK2B/CAMK2D (Phospho-T305)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9UQM7, Q13554, Q13557
- Antigen Synonyms:CAMKB|CaMK-II subunit alpha|CaM kinase II subunit alpha|CAM2|Ca|CAMK2A|CAMK2B|CAMKA|CAMK2|Calcium/calmodulin-dependent protein kinase type II subunit alpha|KIAA0968|Calcium/calmodulin-dependent protein kinase type II subunit beta
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human CaMKIIα/β/δ around the phosphorylation site of T305
- Purification:Antigen affinity purified
- Cat. No.:77698-627
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: CAMK2A/CAMK2B/CAMK2D (phospho-T305)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat