Anti-JUN (Acetyl-K271) Rabbit Polyclonal Antibody
Catalog # 77698-346
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P05412
- Antigen Synonyms:JUNC|AP-1|Transcription factor AP-1|Transcription Factor AP 1|V jun sarcoma virus 17 oncogene homolog|Proto oncogene cJun|Oncogene JUN|cJun|Jun proto oncogene|V jun sarcoma virus 17 oncogene homolog (avian)|JUN_HUMAN|V-jun avian sarcoma virus 17 oncogene homolog|vJun Avian Sarcoma Virus 17 Oncogene Homolog|Jun oncogene|JUN|AP1|Proto oncogene c jun|JUN protein|p39|Enhancer Binding Protein AP1|Jun Activation Domain Binding Protein|Proto-oncogene c-jun|Transcription Factor AP1|AP 1|Activator protein 1|V jun avian sarcoma virus 17 oncogene homolog
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human AP-1 around the acetylation site of K271
- Purification:Antigen affinity purified
- Cat. No.:77698-346
- Packaging:Vial
Specifications
About this item
Type:
Antigen: JUN (acetyl-K271)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat