Anti-Cleaved-COL4A3 (P1426) Rabbit Polyclonal Antibody
Catalog # 77698-269
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Cleaved-COL4A3 (P1426)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q01955
- Antigen Synonyms:COL4A 3|Collagen type IV alpha 3|OTTHUMP00000195044|Tumstatin|Collagen type IV alpha 3 (Goodpasture antigen)|Collagen IV alpha 3 polypeptide|Collagen alpha 3(IV) chain|Alpha 3 type IV collagen|CO4A3_HUMAN|Goodpasture antigen|Alpha3 type IV collagen|Col4a3|Collagen type IV alpha 3 chain
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Tumstatin
- Purification:Antigen affinity purified
- Cat. No.:77698-269
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: Cleaved-COL4A3 (P1426)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey