Anti-Cleaved-C1S (R437) Rabbit Polyclonal Antibody
Catalog # 77698-226
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Cleaved-C1S (R437)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P09871
- Antigen Synonyms:FLJ44757|Complement C1s subcomponent heavy chain|Complement component 1 s subcomponent|Complement C1s subcomponent |C1S_HUMAN|C1S|Complement C1s subcomponent light chain|C1 esterase|Basic proline rich peptide IB 1|Complement component 1 subcomponent s
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human C1s HC
- Purification:Antigen affinity purified
- Cat. No.:77698-226
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: Cleaved-C1S (R437)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human