Anti-DNAJB14 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DNAJB14
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-602
- 77718-603
- Antigen Symbol:
- DNAJB14
- DNAJB14
- Antigen Synonyms:
- member 14|PRO34683|DnaJ homolog subfamily B member 14|DnaJ (Hsp40) homolog|subfamily B|dnajb14|MGC22187|5730496F10Rik|FLJ14281|UNQ9427/PRO34683|DJB14_HUMAN|EGNR9427
- member 14|PRO34683|DnaJ homolog subfamily B member 14|DnaJ (Hsp40) homolog|subfamily B|dnajb14|MGC22187|5730496F10Rik|FLJ14281|UNQ9427/PRO34683|DJB14_HUMAN|EGNR9427
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TBM8
- Q8TBM8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DnaJ homolog subfamily B member 14 protein (1-244AA)
- Recombinant Human DnaJ homolog subfamily B member 14 protein (1-244AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice