Anti-CTCFL Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: CTCFL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-552
- 77718-551
- Antigen Symbol:
- CTCFL
- CTCFL
- Antigen Synonyms:
- HMG 1L1|CTCF-like protein|Putative high mobility group protein B1 like 1|Cancer/testis antigen 27|MGC169106|Zinc finger protein CTCF-T|Transcriptional repressor CTCFL|HMGB1L1|CTCFL_HUMAN|Brother of the regulator of imprinted sites|CCCTC-binding factor|Putative high mobility group protein 1 like 1|CTCF T|Ctcfl|MGC169105|MGC163358|CCCTC binding factor (zinc finger protein) like|dJ579F20.2|CTCF paralog|BORIS like protein|CT27
- HMG 1L1|CTCF-like protein|Putative high mobility group protein B1 like 1|Cancer/testis antigen 27|MGC169106|Zinc finger protein CTCF-T|Transcriptional repressor CTCFL|HMGB1L1|CTCFL_HUMAN|Brother of the regulator of imprinted sites|CCCTC-binding factor|Putative high mobility group protein 1 like 1|CTCF T|Ctcfl|MGC169105|MGC163358|CCCTC binding factor (zinc finger protein) like|dJ579F20.2|CTCF paralog|BORIS like protein|CT27
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NI51
- Q8NI51
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcriptional repressor CTCFL protein (1-270AA)
- Recombinant Human Transcriptional repressor CTCFL protein (1-270AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice