Anti-FANCB Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50-1:200
Type:
Antigen: FANCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-440
- 77718-439
- Antigen Symbol:
- FANCB
- FANCB
- Antigen Synonyms:
- FAAP90|Fancb|Fanconi anemia group B protein|Protein FACB|Fanconi anemia|FA2|FANCB_HUMAN|FAAP95|complementation group B|FAB|Fanconi anemia-associated polypeptide of 95 kDa|Fanconi anemia associated polypeptide of 95 kDa |FACB
- FAAP90|Fancb|Fanconi anemia group B protein|Protein FACB|Fanconi anemia|FA2|FANCB_HUMAN|FAAP95|complementation group B|FAB|Fanconi anemia-associated polypeptide of 95 kDa|Fanconi anemia associated polypeptide of 95 kDa |FACB
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NB91
- Q8NB91
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fanconi anemia group B protein (431-574AA)
- Recombinant Human Fanconi anemia group B protein (431-574AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice