Anti-Abhd11 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Abhd11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77717-549
- 77717-550
- Antigen Symbol:
- Abhd11
- Abhd11
- Antigen Synonyms:
- Wbscr21Protein ABHD11|Williams-Beuren syndrome chromosomal region 21 protein homolog|EC 3.-.-.-|Abhydrolase domain-containing protein 11|Abhd11|Alpha/beta hydrolase domain-containing protein 11
- Wbscr21Protein ABHD11|Williams-Beuren syndrome chromosomal region 21 protein homolog|EC 3.-.-.-|Abhydrolase domain-containing protein 11|Abhd11|Alpha/beta hydrolase domain-containing protein 11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8K4F5
- Q8K4F5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Protein ABHD11 protein (1-307AA)
- Recombinant Mouse Protein ABHD11 protein (1-307AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice