Anti-GPIHBP1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPIHBP1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-440
- 77717-441
- Antigen Symbol:
- GPIHBP1
- GPIHBP1
- Antigen Synonyms:
- High density lipoprotein-binding protein 1|GPI-anchored HDL-binding protein 1|GPI anchored High Density Lipid binding protein 1|Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1|HDBP1_HUMAN|GPI-HBP1|GPI HBP1|GPI anchored HDL binding protein 1|GPIHBP1|Hbp1
- High density lipoprotein-binding protein 1|GPI-anchored HDL-binding protein 1|GPI anchored High Density Lipid binding protein 1|Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1|HDBP1_HUMAN|GPI-HBP1|GPI HBP1|GPI anchored HDL binding protein 1|GPIHBP1|Hbp1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IV16
- Q8IV16
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1 protein (21-151AA)
- Recombinant Human Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1 protein (21-151AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice