Anti-CLEC10A Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CLEC10A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-429
- 77717-428
- Antigen Symbol:
- CLEC10A
- CLEC10A
- Antigen Synonyms:
- CLC10_HUMAN|C-type lectin domain family 10 member A|C-type lectin superfamily member 14|C type (calcium dependent carbohydrate recognition domain) lectin superfamily member 13 (macrophage derived)|C type (calcium dependent carbohydrate recognition domain) lectin superfamily member 14 (macrophage derived)|C type lectin superfamily member 14|Mgl1|CD301A|Macrophage lectin 2|MACROPHAGE GALACTOSE-TYPE C-TYPE|CD301|CLEC10A|Macrophage lectin 2 (calcium dependent)|C type lectin domain family 10|Lectin|MGL|CD301 antigen|HML|Macrophage C type lectin|CLECSF14|member A|HML2|CLECSF13
- CLC10_HUMAN|C-type lectin domain family 10 member A|C-type lectin superfamily member 14|C type (calcium dependent carbohydrate recognition domain) lectin superfamily member 13 (macrophage derived)|C type (calcium dependent carbohydrate recognition domain) lectin superfamily member 14 (macrophage derived)|C type lectin superfamily member 14|Mgl1|CD301A|Macrophage lectin 2|MACROPHAGE GALACTOSE-TYPE C-TYPE|CD301|CLEC10A|Macrophage lectin 2 (calcium dependent)|C type lectin domain family 10|Lectin|MGL|CD301 antigen|HML|Macrophage C type lectin|CLECSF14|member A|HML2|CLECSF13
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IUN9
- Q8IUN9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human C-type lectin domain family 10 member A protein (66-316AA)
- Recombinant Human C-type lectin domain family 10 member A protein (66-316AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice