Anti-HPS6 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HPS6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77716-909
- 77716-908
- Antigen Symbol:
- HPS6
- HPS6
- Antigen Synonyms:
- HPS6_HUMAN|Hermansky Pudlak syndrome 6 protein|Ruby-eye protein homolog|HPS6 biogenesis of lysosomal organelles complex 2 subunit 3|FLJ22501|MGC20522|HPS6|Ru|Ruby eye protein homolog|BLOC2S3|Hermansky-Pudlak syndrome 6 protein
- HPS6_HUMAN|Hermansky Pudlak syndrome 6 protein|Ruby-eye protein homolog|HPS6 biogenesis of lysosomal organelles complex 2 subunit 3|FLJ22501|MGC20522|HPS6|Ru|Ruby eye protein homolog|BLOC2S3|Hermansky-Pudlak syndrome 6 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86YV9
- Q86YV9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hermansky-Pudlak syndrome 6 protein (221-319AA)
- Recombinant Human Hermansky-Pudlak syndrome 6 protein (221-319AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice